2-Chloro-5-fluoro-3-isobutoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77726-500MG 77726-1G 77726-5G
77339-532EA 594.31 USD
77339-532 77358-584 77382-466
2-Chloro-5-fluoro-3-isobutoxybenzaldehyde ≥97%
2-Chloro-5-fluoro-3-isobutoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClFO₂
MW: 230.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767220
CAS Number: 2901105-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR