1-Bromo-2-chloro-5-fluoro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

77707-500MG 77707-1G 77707-5G
77338-278EA 470.71 USD
77338-278 77358-578 77382-462
1-Bromo-2-chloro-5-fluoro-3-isopropoxybenzene ≥97%
1-Bromo-2-chloro-5-fluoro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34767201
CAS Number: 2901065-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR