2-Chloro-3-ethoxy-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

77703-500MG 77703-1G 77703-5G
77340-114EA 761.17 USD
77340-114 77358-576 77382-460
2-Chloro-3-ethoxy-5-fluorobenzoic acid ≥97%
2-Chloro-3-ethoxy-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767197
CAS Number: 2387193-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR