1-(Benzyloxy)-3-bromo-2-chloro-5-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

77700-500MG 77700-1G 77700-5G
77334-360EA 321.36 USD
77334-360 77358-572 77382-456
1-(Benzyloxy)-3-bromo-2-chloro-5-fluorobenzene ≥97%
1-(Benzyloxy)-3-bromo-2-chloro-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34767194
CAS Number: 2901081-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR