1-Bromo-2-chloro-3-ethoxy-5-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

77696-500MG 77696-1G 77696-5G
77338-438EA 487.19 USD
77338-438 77358-568 77382-452
1-Bromo-2-chloro-3-ethoxy-5-fluorobenzene ≥97%
1-Bromo-2-chloro-3-ethoxy-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
MW: 253.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767191
CAS Number: 2386674-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR