2-(Benzyloxy)-1-bromo-3,4-dichlorobenzene ≥95%

Supplier: AOBChem USA

Warning

77649-500MG 77649-1G 77649-5G
77336-124EA 375.95 USD
77336-124 77358-564 77382-450
2-(Benzyloxy)-1-bromo-3,4-dichlorobenzene ≥95%
2-(Benzyloxy)-1-bromo-3,4-dichlorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrCl₂O
MW: 332.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767145
CAS Number: 2901103-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR