1-Bromo-3,4-dichloro-2-isobutoxybenzene ≥95%

Supplier: AOBChem USA

Warning

77648-500MG 77648-1G 77648-5G
77334-358EA 342.99 USD
77334-358 77358-562 77382-448
1-Bromo-3,4-dichloro-2-isobutoxybenzene ≥95%
1-Bromo-3,4-dichloro-2-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD34767144
CAS Number: 2901095-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR