3-Chloro-4-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

77629-500MG 77629-1G 77629-5G
77338-682EA 508.82 USD
77338-682 77358-554 77382-442
3-Chloro-4-isopropylbenzoic acid ≥97%
3-Chloro-4-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD20640283
CAS Number: 161622-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR