(2-Fluoro-5-iodo-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

77626-500MG 77626-1G 77626-5G 77626-25G
77334-356EA 216.3 USD
77334-356 77358-552 77370-350 77391-118
(2-Fluoro-5-iodo-4-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-5-iodo-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIS
MW: 282.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767130
CAS Number: 2383024-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR