4-Fluoro-2-methyl-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

77622-500MG 77622-1G 77622-5G 77622-25G
77334-352EA 156.56 USD
77334-352 77345-628 77370-346 77391-114
4-Fluoro-2-methyl-5-(methylthio)benzoic acid ≥97%
4-Fluoro-2-methyl-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD34767126
CAS Number: 2091538-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR