3-Fluoro-5-methyl-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77615-500MG 77615-1G 77615-5G 77615-25G
77334-348EA 173.04 USD
77334-348 77346-462 77370-342 77391-110
3-Fluoro-5-methyl-2-(methylthio)benzaldehyde ≥97%
3-Fluoro-5-methyl-2-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FOS
MW: 184.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767119
CAS Number: 2090465-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR