3-Fluoro-5-methyl-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

77613-500MG 77613-1G 77613-5G 77613-25G
77334-346EA 99.91 USD
77334-346 77342-890 77370-340 77382-440
3-Fluoro-5-methyl-2-(methylthio)benzoic acid ≥97%
3-Fluoro-5-methyl-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767117
CAS Number: 2901081-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR