(2,6-Difluoro-3-iodo-5-methylphenyl)trimethylsilane ≥97%
Supplier: AOBChem USA
|
Warning
|
77605-500MG
77605-1G
77605-5G
77605-25G
77334-342EA
128.75
USD
77334-342
77344-290
77370-336
77382-438
(2,6-Difluoro-3-iodo-5-methylphenyl)trimethylsilane ≥97%
(2,6-Difluoro-3-iodo-5-methylphenyl)trimethylsilane
Precaución: for Research Use Only
Formula:
C₁₀H₁₃F₂ISi Storage Temperature: Ambient |
MDL Number:
MFCD34767112 CAS Number: 2901099-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...