2-(2,5-Dichloro-4-(methylthio)phenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

77593-500MG 77593-1G 77593-5G
77338-368EA 482.04 USD
77338-368 77358-550 77382-436
2-(2,5-Dichloro-4-(methylthio)phenyl)-1,3-dioxolane ≥95%
2-(2,5-Dichloro-4-(methylthio)phenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂S
MW: 265.1562 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767101
CAS Number: 2901084-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR