2-(4-Chloro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

77592-500MG 77592-1G 77592-5G 77592-25G
77334-340EA 178.19 USD
77334-340 77346-630 77370-334 77391-106
2-(4-Chloro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane ≥97%
2-(4-Chloro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂S
MW: 244.7377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767100
CAS Number: 2901082-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR