2-Bromo-5-chloro-3,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

77489-500MG 77489-1G 77489-5G
77334-336EA 307.97 USD
77334-336 77358-544 77382-432
2-Bromo-5-chloro-3,4'-dimethyl-1,1'-biphenyl ≥97%
2-Bromo-5-chloro-3,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrCl
MW: 295.602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767016
CAS Number: 2901099-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR