5'-Bromo-4'-ethoxy-2',3'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
77481-500MG
77481-1G
77481-5G
77337-254EA
409.94
USD
77337-254
77358-542
77382-430
5'-Bromo-4'-ethoxy-2',3'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%
5'-Bromo-4'-ethoxy-2',3'-difluoro-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₅H₁₁BrF₂O₂ MW: 341.147 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34767012 CAS Number: 2901102-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...