4-(2-Fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

77475-500MG 77475-1G 77475-5G 77475-25G
77334-332EA 245.14 USD
77334-332 77358-540 77370-328 77391-100
4-(2-Fluorophenyl)pyridine ≥97%
4-(2-Fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈FN
Storage Temperature: Ambient
MDL Number: MFCD18416840
CAS Number: 89346-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR