3-Bromo-2-fluoro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

77463-500MG 77463-1G 77463-5G 77463-25G
77334-328EA 184.37 USD
77334-328 77347-340 77370-324 77391-096
3-Bromo-2-fluoro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥95%
3-Bromo-2-fluoro-4'-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD34767009
CAS Number: 2918928-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR