3'-Bromo-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

77457-500MG 77457-1G 77457-5G
77334-324EA 347.11 USD
77334-324 77358-536 77382-426
3'-Bromo-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Bromo-2'-fluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈BrFO
MW: 279.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31596289
CAS Number: 1696744-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR