3-Bromo-2-fluoro-4'-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

77456-500MG 77456-1G 77456-5G 77456-25G
77334-322EA 96.82 USD
77334-322 77342-634 77370-322 77382-424
3-Bromo-2-fluoro-4'-methoxy-1,1'-biphenyl ≥97%
3-Bromo-2-fluoro-4'-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767004
CAS Number: 2901082-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR