5-Bromo-2-iodo-1-isopropoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

77437-500MG 77437-1G 77437-5G
77338-954EA 535.6 USD
77338-954 77358-532 77382-420
5-Bromo-2-iodo-1-isopropoxy-3-methylbenzene ≥95%
5-Bromo-2-iodo-1-isopropoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrIO
MW: 355.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766991
CAS Number: 2901095-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR