5-(3-Bromo-2-ethoxy-5-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

77410-500MG 77410-1G 77410-5G
77340-220EA 840.48 USD
77340-220 77358-530 77382-416
5-(3-Bromo-2-ethoxy-5-methylphenyl)thiazol-2-amine ≥97%
5-(3-Bromo-2-ethoxy-5-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂OS
MW: 313.2134 g/mol
Storage Temperature: Ambient
CAS Number: 2921839-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR