2-(2-Aminothiazol-5-yl)-6-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

77402-500MG 77402-1G
77340-494EA 1274.11 USD
77340-494 77358-528
2-(2-Aminothiazol-5-yl)-6-methoxyphenol ≥95%
2-(2-Aminothiazol-5-yl)-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂S
MW: 222.2636 g/mol
Storage Temperature: Ambient
CAS Number: 2901103-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR