5-(5-Iodo-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

77396-500MG 77396-1G 77396-5G
77340-192EA 826.06 USD
77340-192 77358-524 77382-412
5-(5-Iodo-2-methylphenyl)thiazol-2-amine ≥97%
5-(5-Iodo-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉IN₂S
MW: 316.1613 g/mol
Storage Temperature: Ambient
CAS Number: 2901098-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR