4-Methyl-3-(2-(methylamino)thiazol-5-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

77382-500MG 77382-1G 77382-5G 77382-25G
77334-318EA 195.7 USD
77334-318 77358-522 77370-318 77391-094
4-Methyl-3-(2-(methylamino)thiazol-5-yl)phenol ≥97%
4-Methyl-3-(2-(methylamino)thiazol-5-yl)phenol

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.2907 g/mol
Storage Temperature: Ambient
CAS Number: 2901082-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR