3-(2-Aminothiazol-5-yl)-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

77381-500MG 77381-1G 77381-25G 77381-5G
77334-316EA 301.79 USD
77334-316 77358-520 77391-092 77382-410
3-(2-Aminothiazol-5-yl)-4-methylphenol ≥97%
3-(2-Aminothiazol-5-yl)-4-methylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂OS
MW: 206.2642 g/mol
Storage Temperature: Ambient
CAS Number: 2901066-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR