2,4-Diethoxy-3-fluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
77241-500MG
77241-1G
77241-5G
77339-530EA
594.31
USD
77339-530
77358-518
77382-408
2,4-Diethoxy-3-fluorobenzoic acid ≥97%
2,4-Diethoxy-3-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₃FO₄ MW: 228.217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34766932 CAS Number: 2901085-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...