3-Fluoro-2-formyl-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

77196-500MG 77196-1G 77196-5G 77196-25G
77334-312EA 267.8 USD
77334-312 77358-514 77370-314 77391-088
3-Fluoro-2-formyl-4-propoxybenzoic acid ≥97%
3-Fluoro-2-formyl-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₄
Storage Temperature: Ambient
MDL Number: MFCD34766913
CAS Number: 2918918-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR