1-Bromo-3-fluoro-2-methyl-4-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

77185-500MG 77185-1G 77185-5G 77185-25G
77334-308EA 109.18 USD
77334-308 77343-226 77370-310 77382-404
1-Bromo-3-fluoro-2-methyl-4-propoxybenzene ≥97%
1-Bromo-3-fluoro-2-methyl-4-propoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD34766902
CAS Number: 2901109-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR