3-Fluoro-2-(methylthio)-4-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77181-500MG 77181-1G 77181-5G
77337-778EA 441.87 USD
77337-778 77358-512 77382-402
3-Fluoro-2-(methylthio)-4-propoxybenzaldehyde ≥97%
3-Fluoro-2-(methylthio)-4-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂S
MW: 228.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766898
CAS Number: 2807465-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR