3-Fluoro-2-(methylthio)-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

77179-500MG 77179-1G 77179-25G 77179-5G
77334-306EA 303.85 USD
77334-306 77358-510 77391-084 77382-400
3-Fluoro-2-(methylthio)-4-propoxybenzoic acid ≥97%
3-Fluoro-2-(methylthio)-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃S
Storage Temperature: Ambient
MDL Number: MFCD34766896
CAS Number: 2807443-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR