3,4-Difluoro-5-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77173-500MG 77173-1G 77173-5G
77339-900EA 672.59 USD
77339-900 77358-504 77382-394
3,4-Difluoro-5-(methoxymethyl)benzaldehyde ≥97%
3,4-Difluoro-5-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766890
CAS Number: 2901065-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR