1-Chloro-2-(2,2-diethoxyethoxy)-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

77161-500MG 77161-1G 77161-5G 77161-25G
77334-302EA 178.19 USD
77334-302 77346-698 77370-308 77391-082
1-Chloro-2-(2,2-diethoxyethoxy)-4-fluorobenzene ≥97%
1-Chloro-2-(2,2-diethoxyethoxy)-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34766882
CAS Number: 1998457-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR