2-Chloro-3-ethoxy-4-iodo-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

77084-500MG 77084-1G 77084-5G
77334-300EA 342.99 USD
77334-300 77358-490 77382-376
2-Chloro-3-ethoxy-4-iodo-1-methylbenzene ≥97%
2-Chloro-3-ethoxy-4-iodo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClIO
Storage Temperature: Ambient
MDL Number: MFCD34766830
CAS Number: 2383926-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR