3-Chloro-2-ethoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77081-500MG 77081-1G 77081-5G
77334-296EA 347.11 USD
77334-296 77358-486 77382-372
3-Chloro-2-ethoxy-4-methylbenzaldehyde ≥97%
3-Chloro-2-ethoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34766827
CAS Number: 2386523-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR