3-Chloro-2-ethoxy-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

77079-500MG 77079-1G 77079-5G
77335-592EA 358.44 USD
77335-592 77358-484 77382-370
3-Chloro-2-ethoxy-4-methylbenzoic acid ≥97%
3-Chloro-2-ethoxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.6455 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766825
CAS Number: 2384058-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR