3-Bromo-2-chloro-4-((trimethylsilyl)methyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

76982-500MG 76982-1G 76982-5G 76982-25G
77334-290EA 120.51 USD
77334-290 77343-838 77370-298 77382-362
3-Bromo-2-chloro-4-((trimethylsilyl)methyl)pyridine ≥95%
3-Bromo-2-chloro-4-((trimethylsilyl)methyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₃BrClNSi
MW: 278.6487 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766769
CAS Number: 2901095-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR