3-(3,5-Dibromopyridin-4-yl)-1,1-Difluoropropan-2-one ≥97%

Supplier: AOBChem USA

Warning

76968-500MG 76968-1G 76968-5G
77335-730EA 363.59 USD
77335-730 77358-480 77382-360
3-(3,5-Dibromopyridin-4-yl)-1,1-Difluoropropan-2-one ≥97%
3-(3,5-Dibromopyridin-4-yl)-1,1-Difluoropropan-2-one

Precaución: for Research Use Only

Formula: C₈H₅Br₂F₂NO
MW: 328.9362 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766765
CAS Number: 2901102-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR