2-(6-Bromo-3-methoxypyridin-2-yl)acetic acid ≥95%

Supplier: AOBChem USA

Warning

76951-500MG 76951-1G 76951-5G
77339-698EA 614.91 USD
77339-698 77358-478 77382-358
2-(6-Bromo-3-methoxypyridin-2-yl)acetic acid ≥95%
2-(6-Bromo-3-methoxypyridin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD28741640
CAS Number: 1807214-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR