1,1-Difluoro-3-(pyridin-2-yl)propan-2-one ≥97%

Supplier: AOBChem USA

Warning

76923-500MG 76923-1G 76923-25G 76923-5G
77334-288EA 284.28 USD
77334-288 77358-476 77391-080 77382-356
1,1-Difluoro-3-(pyridin-2-yl)propan-2-one ≥97%
1,1-Difluoro-3-(pyridin-2-yl)propan-2-one

Precaución: for Research Use Only

Formula: C₈H₇F₂NO
MW: 171.1440 g/mol
Storage Temperature: Ambient
CAS Number: 1566148-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR