2-Fluoro-6-iodo-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

76863-500MG 76863-1G 76863-5G 76863-25G
77334-286EA 264.71 USD
77334-286 77358-474 77370-296 77391-078
2-Fluoro-6-iodo-3-methoxyphenol ≥97%
2-Fluoro-6-iodo-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34766756
CAS Number: 2386767-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR