3-Fluoro-2-hydroxy-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76858-500MG 76858-1G 76858-5G
77339-572EA 597.4 USD
77339-572 77358-472 77382-354
3-Fluoro-2-hydroxy-4-methoxybenzaldehyde ≥97%
3-Fluoro-2-hydroxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23705522
CAS Number: 1427397-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR