3-Fluoro-2-hydroxy-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76855-500MG 76855-1G 76855-5G 76855-25G
77334-284EA 141.11 USD
77334-284 77344-934 77370-294 77391-076
3-Fluoro-2-hydroxy-4-methoxybenzoic acid ≥97%
3-Fluoro-2-hydroxy-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₄
MW: 186.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28554880
CAS Number: 1784962-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR