3-Fluoro-2-isopropoxy-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76836-500MG 76836-1G 76836-5G 76836-25G
77334-280EA 196.73 USD
77334-280 77358-468 77370-292 77391-072
3-Fluoro-2-isopropoxy-4-methoxybenzoic acid ≥97%
3-Fluoro-2-isopropoxy-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
MW: 228.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766733
CAS Number: 2901102-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR