3-Fluoro-2-isobutoxy-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76830-500MG 76830-1G 76830-5G
77337-530EA 416.12 USD
77337-530 77358-464 77382-350
3-Fluoro-2-isobutoxy-4-methoxybenzaldehyde ≥97%
3-Fluoro-2-isobutoxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
MW: 226.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766727
CAS Number: 2901107-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR