2-Ethoxy-3-fluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76819-500MG 76819-1G 76819-5G 76819-25G
77334-270EA 264.71 USD
77334-270 77358-458 77370-282 77391-064
2-Ethoxy-3-fluoro-4-methoxybenzoic acid ≥97%
2-Ethoxy-3-fluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
MW: 214.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766716
CAS Number: 2386826-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR