(3-Fluoro-2,4-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

76816-500MG 76816-1G 76816-5G
77336-966EA 400.67 USD
77336-966 77358-456 77382-346
(3-Fluoro-2,4-dimethoxyphenyl)(methyl)sulfane ≥97%
(3-Fluoro-2,4-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD28793148
CAS Number: 1804420-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR