2-(Benzyloxy)-1-bromo-3-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

76812-500MG 76812-1G 76812-5G 76812-25G
77334-268EA 141.11 USD
77334-268 77344-932 77370-280 77391-062
2-(Benzyloxy)-1-bromo-3-fluoro-4-methoxybenzene ≥97%
2-(Benzyloxy)-1-bromo-3-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766712
CAS Number: 2901095-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR