1-Bromo-2-ethoxy-3-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

76810-500MG 76810-1G 76810-5G 76810-25G
77334-264EA 116.39 USD
77334-264 77343-466 77370-276 77382-342
1-Bromo-2-ethoxy-3-fluoro-4-methoxybenzene ≥97%
1-Bromo-2-ethoxy-3-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766710
CAS Number: 2383779-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR